Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139295.3 | NP_647456.1 | 2, 3 | 282 | ||
NM_176808.5 | NP_789778.1 | 3, 4 | 507 | |||
GenBank mRNA | AF475283.1 | - | 2, 3 | 250 | ||
AK033139.1 | - | 2, 3 | 216 | |||
AK075781.1 | - | 3, 4 | 440 | |||
AK077139.1 | - | 2, 3 | 233 | |||
AK077631.1 | - | 2, 3 | 216 | |||
AK141599.1 | - | 3, 4 | 465 | |||
AK151803.1 | - | 2, 3 | 271 | |||
AK206943.1 | - | 1, 2 | 161 | |||
BC003996.1 | - | 2, 3 | 209 | 3492270 | ||
GQ896355.1 | - | 1, 2 | 153 | |||
GQ896356.1 | - | 1, 2 | 153 |