Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145147.5 | NP_660129.2 | 7 | 925 | ||
GenBank mRNA | AJ293619.1 | - | 6 | 555 | ||
AK082192.1 | - | 4 | 801 | |||
AK144812.1 | - | 7 | 926 | |||
AK152225.1 | - | 7 | 926 | |||
AK152972.1 | - | 7 | 926 | |||
AK159462.1 | - | 7 | 898 | |||
BC027821.1 | - | 4 | 456 | 5321725 |