Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146377.1 | NP_666489.1 | 1 | 886 | ||
GenBank mRNA | BC132433.1 | - | 1 | 912 | 40130710 | |
BC145997.1 | - | 1 | 912 | 40131300 |