Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146704.1 | NP_666915.1 | 1 | 466 | ||
GenBank mRNA | BC119061.1 | - | 1 | 549 | 8733810 | |
BC137821.1 | - | 1 | 549 | 8860840 |