Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177632.3 | NP_808300.1 | 1 | 1056 | ||
GenBank mRNA | AK083036.1 | - | 1 | 1056 | ||
BC099947.1 | - | 1 | 1069 | 30088416 | ||
BC138882.1 | - | 1 | 670 | 8861904 | ||
BC138884.1 | - | 1 | 646 | 8861906 |