Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001697.2 | NP_001688.1 | 2, 3 | 185 | ||
GenBank mRNA | AK124187.1 | - | 1, 2 | 1326 | ||
AK222608.1 | - | 2, 3 | 117 | |||
AK222962.1 | - | 2, 3 | 112 | |||
AK311796.1 | - | 2, 3 | 112 | |||
AV734214.1 | - | 2, 3 | 185 | |||
BC021233.2 | - | 2, 3 | 98 | 4126036 | ||
BC022865.1 | - | 2, 3 | 116 | 4103001 | ||
BT019836.1 | - | 2, 3 | 94 | |||
CD174837.1 | - | 2, 3 | 155 | |||
CR456822.1 | - | 2, 3 | 94 | |||
DQ656021.2 | - | 2, 3 | 120 | |||
DQ656022.2 | - | 2, 3 | 132 | |||
X83218.1 | - | 2, 3 | 130 |