Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148947.1 | NP_683749.1 | 2 | 179 | ||
GenBank mRNA | AJ315552.1 | - | 2 | 179 | ||
BC111447.1 | - | 2 | 178 | |||
BC145966.1 | - | 2 | 200 | 40131269 | ||
BC145968.1 | - | 2 | 200 | 40131271 |