Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bricd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175682.3 | NP_783613.1 | 4 | 628 | ||
GenBank mRNA | AK079437.1 | - | 4 | 629 | ||
BC096398.1 | - | 4 | 389 | |||
BC119389.1 | - | 5 | 414 | 8734138 | ||
BC120670.1 | - | 5 | 414 | 40129593 | ||
BC145010.1 | - | 5 | 414 | 9053558 |