Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001711.5 | NP_001702.1 | 3 | 565 | ||
XM_017029724.1 | XP_016885213.1 | 2 | 344 | |||
GenBank mRNA | AK093023.1 | - | 3 | 399 | ||
AK094059.1 | - | 4 | 374 | |||
AK222484.1 | - | 3 | 474 | |||
AK223302.1 | - | 3 | 445 | |||
AK291955.1 | - | 3 | 476 | |||
AK293289.1 | - | 3 | 400 | |||
AK298756.1 | - | 2 | 274 | |||
AU142113.1 | - | 3 | 501 | |||
BC002416.2 | - | 3 | 458 | 3162633 | ||
BC004244.1 | - | 3 | 468 | 3503374 | ||
BT007323.1 | - | 2 | 329 | |||
DC419932.1 | - | 3 | 545 | |||
J04599.1 | - | 3 | 449 |