Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290491.1 | NP_001277420.1 | 5 | 846 | ||
NM_001290492.1 | NP_001277421.1 | 5 | 858 | |||
NM_001290493.1 | NP_001277422.1 | 4 | 463 | |||
NM_177370.4 | NP_796344.1 | 5 | 846 | |||
GenBank mRNA | AK053223.1 | - | 5 | 681 | ||
AK077457.1 | - | 5 | 682 | |||
AK156451.1 | - | 5 | 700 | |||
AK212977.1 | - | 1 | 99 | |||
AK218833.1 | - | 1 | 99 | |||
BC062155.1 | - | 3 | 561 | |||
BC096693.1 | - | 4 | 472 | 5255059 | ||
BC132459.1 | - | 4 | 511 | 40130736 | ||
BC138479.1 | - | 4 | 511 | 8861499 | ||
BC145287.1 | - | 4 | 523 | 9053841 | ||
BC145289.1 | - | 4 | 511 | 9053843 | ||
BC145290.1 | - | 4 | 511 |