Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007526.4 | NP_031552.2 | 2 | 523 | ||
GenBank mRNA | AB221538.1 | - | 1 | 121 | ||
AJ297677.1 | - | 2 | 550 | |||
BC119047.1 | - | 1 | 202 | |||
BC119049.1 | - | 1 | 202 | |||
Y07960.1 | - | 2 | 265 |