Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007923.2 | NP_031949.2 | 3 | 576 | ||
GenBank mRNA | AK054075.1 | - | 3 | 868 | ||
AK156537.1 | - | 3 | 574 | |||
BC144844.1 | - | 3 | 326 | 9053392 | ||
BC145795.1 | - | 3 | 326 | 40131098 | ||
FN434116.1 | - | 2 | 173 | |||
FN434118.1 | - | 2 | 173 | |||
Z36885.1 | - | 4 | 521 |