Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139051.3 | NP_620639.1 | 2 | 252 | ||
GenBank mRNA | AB000490.1 | - | 2 | 296 | ||
AF511594.1 | - | 2 | 82 | |||
AK134378.1 | - | 2 | 253 | |||
AK144376.1 | - | 2 | 213 | |||
AK156911.1 | - | 2 | 220 | |||
BC110476.1 | - | 2 | 195 | 40047261 | ||
BC110477.1 | - | 2 | 195 | 40047262 | ||
D10584.1 | - | 1 | 482 |