Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015728.1 | - | 2 | 464 | ||
BC139217.1 | - | 2 | 333 | 8862239 | ||
BC139218.1 | - | 2 | 333 | 8862240 | ||
BC145578.1 | - | 2 | 330 | 9054144 | ||
BG082217.2 | - | 2 | 96 | |||
L76741.1 | - | 2 | 73 |