Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasl2-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009028.2 | NP_033054.1 | 1 | 98 | ||
GenBank mRNA | AK006350.1 | - | 1 | 99 | ||
BC049619.1 | - | 1 | 101 | |||
BC061180.1 | - | 1 | 98 | 6771852 | ||
BC099553.1 | - | 1 | 98 | 30917698 | ||
BC138995.1 | - | 1 | 100 | 8862017 | ||
BC145722.1 | - | 1 | 100 | 40131025 | ||
L32752.1 | - | 1 | 114 |