Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010887.2 | NP_035017.2 | 2 | 153 | ||
GenBank mRNA | AF124785.1 | - | 2 | 103 | ||
AK011124.1 | - | 2 | 141 | |||
AK020552.1 | - | 2 | 109 | |||
BC004618.1 | - | 2 | 111 | 3585543 | ||
CJ073528.1 | - | 2 | 152 |