Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011186.1 | NP_035316.1 | 1, 2 | 210 | ||
GenBank mRNA | AB003304.1 | - | 1, 2 | 213 | ||
AF060091.1 | - | 1, 2 | 210 | |||
AK010441.1 | - | 1, 2 | 230 | |||
AK088367.1 | - | 1, 2 | 223 | |||
AK133839.1 | - | 1, 2 | 231 | |||
AK219104.1 | - | 1, 2 | 38 | |||
BC012246.1 | - | 1, 2 | 218 | 4167355 | ||
BC106144.1 | - | 1, 2 | 213 | 4953887 |