Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011297.2 | NP_035427.2 | 2, 3 | 149 | ||
NM_207634.1 | NP_997517.1 | 2, 3 | 149 | |||
NM_207635.1 | NP_997518.1 | 2, 3 | 149 | |||
GenBank mRNA | AK002495.1 | - | 2, 3 | 113 | ||
AK002986.1 | - | 512 | ||||
AK005358.1 | - | 2, 3 | 113 | |||
AK007776.1 | - | 2, 3 | 113 | |||
AK008386.1 | - | 2, 3 | 113 | |||
AK009048.1 | - | 2, 3 | 113 | |||
AK010856.1 | - | 2, 3 | 113 | |||
AK012537.1 | - | 2, 3 | 100 | |||
AK018704.1 | - | 2, 3 | 113 | |||
AK049387.1 | - | 109 | ||||
AK052480.1 | - | 2, 3 | 113 | |||
AK135247.1 | - | 2, 3 | 102 | |||
AK151111.1 | - | 2, 3 | 111 | |||
AK167874.1 | - | 1, 2 | 84 | |||
BC058817.1 | - | 2, 3 | 109 | 30058168 | ||
BC081457.1 | - | 2, 3 | 95 | 5712639 | ||
BC086882.1 | - | 2, 3 | 112 | 30120790 | ||
BC091748.1 | - | 2, 3 | 93 | 2812455 | ||
BC092287.1 | - | 1, 2 | 80 | |||
BY311117.1 | - | 2, 3 | 203 | |||
X60289.1 | - | 2, 3 | 104 |