Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011731.3 | NP_035861.2 | 1 | 309 | ||
GenBank mRNA | AF075261.1 | - | 1 | 133 | ||
AK143893.1 | - | 1 | 279 | |||
AK144038.1 | - | 1 | 310 | |||
AK163340.1 | - | 1 | 72 | |||
AK164706.1 | - | 1 | 154 | |||
AK164713.1 | - | 1 | 144 | |||
BC013484.1 | - | 1 | 220 | 4242168 |