Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr0b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011850.2 | NP_035980.1 | 1 | 247 | ||
GenBank mRNA | AK004930.1 | - | 1 | 257 | ||
AY360323.1 | - | 1 | 212 | |||
AY360324.1 | - | 1 | 212 | |||
BC019540.1 | - | 1 | 247 | 4193866 |