Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF480417.1 | - | 3 | 536 | ||
BC018470.1 | - | 520 | 3493001 | |||
BC043339.1 | - | 3 | 551 | 5004267 | ||
X58077.1 | - | 463 |