Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164047.1 | NP_001157519.1 | 1, 2 | 297 | ||
NM_001164048.1 | NP_001157520.1 | 2, 3 | 157 | |||
NM_013593.3 | NP_038621.2 | 2, 3 | 245 | |||
GenBank mRNA | AA271284.1 | - | 2, 3 | 157 | ||
AK137456.1 | - | 1, 2 | 152 | |||
BC025172.1 | - | 2, 3 | 257 | 3979003 |