Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001739.1 | NP_001730.1 | 2 | 373 | ||
XM_005256134.4 | XP_005256191.1 | 2 | 442 | |||
XM_011523309.2 | XP_011521611.1 | 2 | 442 | |||
XM_011523310.2 | XP_011521612.1 | 2 | 442 | |||
XM_017023646.1 | XP_016879135.1 | 2 | 442 | |||
GenBank mRNA | BC137405.1 | - | 2 | 369 | 9021403 | |
BC137411.1 | - | 2 | 369 | 9021409 | ||
L19297.1 | - | 2 | 372 |