Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019488.4 | NP_062361.1 | 6, 7 | 915 | ||
GenBank mRNA | AF232061.2 | - | 6, 7 | 872 | ||
AJ245936.1 | - | 6, 7 | 892 | |||
AK081806.1 | - | 6, 7 | 932 | |||
AK155792.1 | - | 6, 7 | 944 | |||
AK170319.1 | - | 6, 7 | 928 | |||
AY856043.1 | - | 6, 7 | 876 | |||
BC021758.1 | - | 2, 3 | 241 | |||
BC090993.1 | - | 6, 7 | 915 | 6751763 | ||
Y17802.1 | - | 6, 7 | 897 |