Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305624.1 | NP_001292553.1 | 4 | 327 | ||
NM_001305625.1 | NP_001292554.1 | 3 | 186 | |||
NM_001305626.1 | NP_001292555.1 | 2 | 3541 | |||
NM_001743.5 | NP_001734.1 | 3 | 281 | |||
GenBank mRNA | AK055130.1 | - | 2 | 1998 | ||
AK315694.1 | - | 3 | 169 | |||
BC003354.1 | - | 3 | 157 | 2899899 | ||
BC006464.1 | - | 3 | 159 | 3542044 | ||
BC008437.1 | - | 3 | 175 | 4092978 | ||
BC017385.1 | - | 3 | 173 | 3681165 | ||
BC018677.1 | - | 3 | 174 | 4656551 | ||
BC026065.1 | - | 3 | 173 | 3683323 | ||
BG563041.1 | - | 4 | 221 | |||
BP370358.1 | - | 3 | 261 | |||
BT009916.1 | - | 2 | 101 | |||
CR541990.1 | - | 2 | 101 | |||
CR542021.1 | - | 2 | 101 | |||
D45887.1 | - | 3 | 169 | |||
HY022313.1 | - | 3 | 281 | |||
HY108355.1 | - | 3 | 187 |