Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023456.2 | NP_075945.1 | 2 | 208 | ||
GenBank mRNA | AF273768.1 | - | 1 | 135 | ||
AK002982.1 | - | 2 | 221 | |||
BC043012.1 | - | 2 | 208 | 5683102 |