Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039392.2 | NP_001034481.1 | 1 | 306 | ||
NM_001190327.1 | NP_001177256.1 | 3 | 317 | |||
NM_025284.4 | NP_079560.1 | 2 | 245 | |||
GenBank mRNA | AK003178.1 | - | 2 | 162 | ||
AK005512.1 | - | 2 | 162 | |||
AK008557.1 | - | 2 | 162 | |||
AK012361.1 | - | 2 | 162 | |||
AK019212.1 | - | 2 | 162 | |||
AK019232.1 | - | 2 | 163 | |||
AK131711.1 | - | 2 | 162 | |||
AK161987.1 | - | 2 | 162 | |||
BC037153.1 | - | 2 | 153 | |||
BC038926.1 | - | 2 | 163 | |||
BC048070.1 | - | 2 | 141 | |||
BC059777.1 | - | 2 | 149 | |||
BC070416.1 | - | 2 | 140 | |||
BC080312.1 | - | 2 | 143 | |||
BC089326.1 | - | 2 | 121 | |||
BC089327.1 | - | 1 | 101 | |||
BC093587.1 | - | 2 | 147 | |||
BC094284.1 | - | 2 | 135 | |||
BC099374.1 | - | 2 | 144 | |||
BC145877.1 | - | 2 | 122 | 40131180 | ||
BC145879.1 | - | 2 | 122 | 40131182 | ||
CA466695.1 | - | 309 | ||||
CB273987.1 | - | 1 | 252 | |||
CJ042349.1 | - | 1 | 376 | |||
CJ042420.1 | - | 1 | 248 | |||
Z48496.1 | - | 1 | 239 |