Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170911.1 | NP_001164382.1 | 3 | 420 | ||
NM_025385.3 | NP_079661.1 | 4 | 538 | |||
GenBank mRNA | AK002430.1 | - | 3 | 388 | ||
AK003850.1 | - | 4 | 535 | |||
AK008611.1 | - | 2 | 360 | |||
AK008894.1 | - | 4 | 498 | |||
AK154583.1 | - | 4 | 495 | |||
AK181565.1 | - | 1 | 195 | |||
AK219155.1 | - | 1 | 155 | |||
BC016234.1 | - | 3 | 684 | 4503179 |