Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psenen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025498.2 | NP_079774.1 | 3 | 368 | ||
GenBank mRNA | AK002440.1 | - | 3 | 316 | ||
AK006284.1 | - | 3 | 333 | |||
AK007621.1 | - | 3 | 331 | |||
BC024347.1 | - | 3 | 368 | 4984284 | ||
BC081443.1 | - | 3 | 370 | 6814799 |