Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025797.3 | NP_080073.1 | 1, 2 | 190 | ||
GenBank mRNA | AK002426.1 | - | 1, 2 | 191 | ||
AK013210.1 | - | 1, 2 | 191 | |||
AK132102.1 | - | 1, 2 | 213 | |||
BC024341.1 | - | 1, 2 | 195 | 4973059 |