Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026308.2 | NP_080584.1 | 4 | 280 | ||
GenBank mRNA | AJ504717.1 | - | 4 | 287 | ||
AK002780.1 | - | 4 | 281 | |||
AK003455.1 | - | 4 | 278 | |||
AY251016.1 | - | 4 | 302 | |||
AY251017.1 | - | 3 | 201 | |||
BC028491.1 | - | 4 | 290 | 1497187 |