Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026467.3 | NP_080743.1 | 2 | 143 | ||
GenBank mRNA | AK007689.1 | - | 2 | 144 | ||
AK011262.1 | - | 2 | 162 | |||
AK151083.1 | - | 2 | 162 | |||
BC058115.1 | - | 2 | 144 | 6407738 |