Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026911.3 | NP_081187.2 | 3 | 566 | ||
GenBank mRNA | AK007336.1 | - | 3 | 264 | ||
AK082928.1 | - | 3 | 265 | |||
AK088658.1 | - | 2 | 349 | |||
BC020050.1 | - | 468 | ||||
BC034270.2 | - | 3 | 441 | |||
BC049895.1 | - | 3 | 257 | 3991764 |