Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001761.2 | NP_001752.2 | 3 | 352 | ||
XM_017023822.1 | XP_016879311.1 | 3 | 512 | |||
XM_017023823.1 | XP_016879312.1 | 3 | 512 | |||
GenBank mRNA | AK313371.1 | - | 3 | 305 | ||
BC012349.1 | - | 3 | 276 | 4634116 | ||
HY027359.1 | - | 3 | 355 | |||
U17105.1 | - | 3 | 277 | |||
Z36714.1 | - | 3 | 307 |