Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029272.3 | NP_083548.1 | 3 | 109 | ||
GenBank mRNA | AK003132.1 | - | 3 | 110 | ||
BC013503.1 | - | 3 | 86 | 4223916 |