Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289554.1 | NP_001276483.1 | 6 | 645 | ||
NM_001289555.1 | NP_001276484.1 | 6 | 642 | |||
NM_001289556.1 | NP_001276485.1 | 5 | 486 | |||
NM_029659.4 | NP_083935.3 | 6 | 652 | |||
GenBank mRNA | AK005599.1 | - | 6 | 643 | ||
AK005858.1 | - | 5 | 393 | |||
AK161310.1 | - | 6 | 653 | |||
BC045148.1 | - | 6 | 582 | 6307739 |