Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ear6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053111.2 | NP_444341.1 | 2 | 171 | ||
GenBank mRNA | AK157836.1 | - | 2 | 168 | ||
AY316150.1 | - | 2 | 171 | |||
BC147690.1 | - | 1 | 403 | 9056663 | ||
BC147697.1 | - | 1 | 403 | 9056670 |