Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134138.1 | NP_598899.1 | 2 | 152 | ||
GenBank mRNA | AB031387.1 | - | 2 | 152 | ||
AK006069.1 | - | 2 | 153 | |||
AK077569.1 | - | 2 | 139 | |||
AK135578.1 | - | 2 | 144 | |||
AK167840.1 | - | 2 | 393 | |||
BC016606.1 | - | 2 | 131 | 2646038 |