Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001769.3 | NP_001760.1 | 2, 3 | 367 | ||
XM_005253814.3 | XP_005253871.1 | 2, 3 | 364 | |||
XM_011521040.1 | XP_011519342.1 | 2, 3 | 346 | |||
GenBank mRNA | AB079244.1 | - | 2, 3 | 288 | ||
AK296332.1 | - | 2, 3 | 310 | |||
AK296894.1 | - | 2, 3 | 283 | |||
AK308700.1 | - | 2, 3 | 314 | |||
AK311020.1 | - | 2, 3 | 346 | |||
AK315219.1 | - | 2, 3 | 316 | |||
AY423720.1 | - | 2, 3 | 200 | |||
AY966455.1 | - | 2, 3 | 294 | |||
BC011988.1 | - | 2, 3 | 234 | 3860667 | ||
BG291377.1 | - | 2, 3 | 369 | |||
BM763066.1 | - | 2, 3 | 367 | |||
L34068.1 | - | 2, 3 | 183 | |||
M38690.1 | - | 2, 3 | 234 | |||
X60111.1 | - | 2, 3 | 294 |