Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138743.2 | NP_620082.1 | 3 | 254 | ||
GenBank mRNA | AK004538.1 | - | 3 | 254 | ||
AY033901.1 | - | 3 | 249 | |||
BC005668.1 | - | 3 | 238 | 3152735 | ||
BM932344.1 | - | 3 | 238 |