Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Porcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB036746.1 | - | 2 | 265 | ||
AB036747.1 | - | 2 | 265 | |||
AB036748.1 | - | 2 | 265 | |||
AB036749.1 | - | 2 | 265 | |||
AK010405.1 | - | 2 | 243 | |||
BC027292.1 | - | 2 | 217 | |||
BC032284.1 | - | 2 | 235 | 5362485 | ||
BF462782.1 | - | 2 | 258 | |||
BQ556448.1 | - | 1 | 82 |