Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040031.1 | NP_001035120.1 | 3 | 288 | ||
NM_001774.2 | NP_001765.1 | 3 | 322 | |||
XM_005259435.3 | XP_005259492.1 | 3 | 324 | |||
XM_005259436.3 | XP_005259493.1 | 3 | 320 | |||
XM_011527542.2 | XP_011525844.1 | 3 | 285 | |||
XM_011527543.2 | XP_011525845.1 | 3 | 390 | |||
XM_011527544.2 | XP_011525846.1 | 3 | 359 | |||
XM_017027513.1 | XP_016883002.1 | 3 | 320 | |||
GenBank mRNA | AK058093.1 | - | 4 | 370 | ||
AK301016.1 | - | 3 | 312 | |||
AK301328.1 | - | 3 | 315 | |||
AK316348.1 | - | 3 | 212 | |||
AL833693.1 | - | 2 | 1366 | |||
BC106752.1 | - | 3 | 239 | 40023348 | ||
BC110379.1 | - | 3 | 274 | 5752290 | ||
BP309557.1 | - | 3 | 342 | |||
BT019851.1 | - | 3 | 201 | |||
CR541877.1 | - | 3 | 201 | |||
X14046.1 | - | 3 | 264 |