Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 11, 12 | 1563 | ||
XM_005273201.1 | XP_005273258.1 | 11, 12 | 1459 | |||
XM_011544238.1 | XP_011542540.1 | 12, 13 | 1655 | |||
XM_011544239.2 | XP_011542541.1 | 13, 14 | 1678 | |||
XM_011544240.2 | XP_011542542.1 | 11, 12 | 1472 | |||
XM_011544241.2 | XP_011542543.1 | 11, 12 | 1434 | |||
XM_017001852.1 | XP_016857341.1 | 11, 12 | 1653 | |||
GenBank mRNA | AB073905.1 | - | 11, 12 | 1486 | ||
AF218003.1 | - | 3, 4 | 578 | |||
AJ278126.1 | - | 7, 8 | 781 | |||
AK074693.1 | - | 11, 12 | 1506 | |||
AK090494.1 | - | 11, 12 | 1300 | |||
AK092784.1 | - | 11, 12 | 1286 | |||
AK126200.1 | - | 4, 5 | 2251 | |||
AK297735.1 | - | 8, 9 | 842 | |||
AK300537.1 | - | 7, 8 | 652 | |||
AK301012.1 | - | 13, 14 | 1484 | |||
BC005171.2 | - | 11, 12 | 1512 | 3604948 | ||
BX648860.1 | - | 11, 12 | 1384 |