Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147103.2 | NP_667314.2 | 1 | 80 | ||
GenBank mRNA | BC106981.1 | - | 1 | 80 | 40043127 | |
BC106982.1 | - | 1 | 80 | 40043129 |