Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175102.4 | NP_780311.2 | 1 | 265 | ||
GenBank mRNA | AK003475.1 | - | 1 | 234 | ||
AK009874.1 | - | 1 | 228 | |||
AK010348.1 | - | 1 | 240 | |||
BB652104.1 | - | 1 | 265 | |||
BC006603.1 | - | 1 | 261 | 3965951 |