Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178933.2 | NP_849264.1 | 2, 3 | 153 | ||
GenBank mRNA | AK002463.1 | - | 2, 3 | 284 | ||
AK004188.1 | - | 2, 3 | 336 | |||
AY279396.1 | - | 2, 3 | 153 | |||
BC115606.1 | - | 1, 2 | 132 | 40102690 | ||
BC115607.1 | - | 1, 2 | 132 | 40102691 |