Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244580.1 | NP_001231509.1 | 4, 5 | 463 | ||
NM_003496.3 | NP_003487.1 | 4, 5 | 463 | |||
GenBank mRNA | AF076974.1 | - | 4, 5 | 463 | ||
AF110377.1 | - | 4, 5 | 383 |