Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004444.4 | NP_004435.3 | 7 | 1903 | ||
XM_017011816.1 | XP_016867305.1 | 7 | 1560 | |||
GenBank mRNA | AB209644.1 | - | 6 | 2613 | ||
AK301224.1 | - | 2 | 210 | |||
AY056048.1 | - | 7 | 1497 | |||
BC004264.1 | - | 4 | 988 | |||
BC052804.1 | - | 7 | 1580 | 6584384 | ||
EU826608.1 | - | 7 | 1412 | |||
U07695.1 | - | 7 | 1497 |