Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010938.1 | NP_001010938.1 | 3 | 543 | ||
NM_001308046.1 | NP_001294975.1 | 3 | 601 | |||
NM_005781.4 | NP_005772.3 | 3 | 752 | |||
XM_005269268.3 | XP_005269325.1 | 3 | 543 | |||
XM_005269270.3 | XP_005269327.1 | 3 | 391 | |||
XM_005269275.3 | XP_005269332.1 | 3 | 565 | |||
XM_011512317.2 | XP_011510619.1 | 5 | 1110 | |||
XM_011512318.2 | XP_011510620.2 | 3 | 482 | |||
XM_011512320.1 | XP_011510622.1 | 3 | 265 | |||
XM_017005508.1 | XP_016860997.1 | 3 | 482 | |||
XM_017005509.1 | XP_016860998.1 | 3 | 482 | |||
XM_017005510.1 | XP_016860999.1 | 3 | 482 | |||
GenBank mRNA | AK131539.1 | - | 3 | 543 | ||
AK225786.1 | - | 3 | 548 | |||
AK309332.1 | - | 3 | 379 | |||
BC008884.2 | - | 3 | 398 | |||
BC020389.1 | - | 3 | 592 | |||
BC028164.1 | - | 3 | 266 | |||
L13738.2 | - | 3 | 827 |