Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319245.1 | NP_001306174.1 | 1, 2 | 468 | ||
NM_001324320.1 | NP_001311249.1 | 1, 2 | 474 | |||
NM_015112.2 | NP_055927.2 | 1, 2 | 468 | |||
XM_011541059.2 | XP_011539361.1 | 1, 2 | 486 | |||
XM_011541061.2 | XP_011539363.1 | 1, 2 | 486 | |||
XM_011541062.2 | XP_011539364.1 | 1, 2 | 486 | |||
XM_011541069.2 | XP_011539371.1 | 1, 2 | 486 | |||
GenBank mRNA | AB047005.1 | - | 1, 2 | 468 | ||
BC065499.1 | - | 1, 2 | 442 | 5499830 |